Benzil ≥99.0%

Supplier: TCI America
Warning

B0050-25G B0050-500G
TCB0050-025GEA 56.52 CAD
TCB0050-025G TCB0050-500G
Benzil ≥99.0%
Benzil

CAS Number: 134-81-6
MDL Number: MFCD00003080
Molecular Formula: C14H10O2
Molecular Weight: 210.23
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 348
Melting point (°C): 96
Flash Point (°C): 180


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Dibenzoyl, Diphenylethanedione, Diphenylglyoxal

Formula: (C₆H₅)C(O)C(O)(C₆H₅)
Boiling Pt: 348 °C
Melting Pt: 96 °C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003080
CAS Number: 134-81-6
EINECS: 205-157-0
Merck Index: 13,01078
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a18e91309875edb4d9a8c6319d11f7cc04690a7&vid=4a18e91309875edb4d9a8c6319d11f7cc04690a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR