Trimesic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid
B0043-500G
B0043-25G
B0043-100G
TCB0043-500GEA
356.38
CAD
TCB0043-500G
TCB0043-025G
TCB0043-100G
Trimesic acid ≥98.0%
Trimesic acid
CAS Number: 554-95-0
MDL Number: MFCD00002517
Molecular Formula: C9H6O6
Molecular Weight: 210.14
Purity/Analysis Method:
Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: Trimesic Acid, BTC
Formula:
C₉H₆O₆ MW: 210.14 g/mol Melting Pt: 345…350 °C Flash Pt: 380 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002517 CAS Number: 554-95-0 EINECS: 209-077-7 |
|
Specification Test Results
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...