Pyromellitic dianhydride ≥98.0%

Supplier: TCI America

Synonyms: 1,2,4,5-Benzenetetracarboxylic anhydride

B0040-25G B0040-500G B0040-100G
TCB0040-025GEA 30.67 CAD
TCB0040-025G TCB0040-500G TCB0040-100G
Pyromellitic dianhydride ≥98.0%
Pyromellitic dianhydride

CAS Number: 89-32-7
MDL Number: MFCD00005005
Molecular Formula: C10H2O6
Molecular Weight: 218.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 400
Melting point (°C): 286
Flash Point (°C): 167


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Benzene-1,2,4,5-tetracarboxylic Dianhydride

Formula: C₁₀H₂O₆
MW: 218.12 g/mol
Boiling Pt: 400 °C
Melting Pt: 286 °C
Flash Pt: 167 °C
MDL Number: MFCD00005005
CAS Number: 89-32-7
EINECS: 201-898-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=774edc02076f4c992f83099ac9c0ca8aef7dd30a&vid=774edc02076f4c992f83099ac9c0ca8aef7dd30a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR