Sodium benzenesulfinate dihydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B0027-25G
B0027-500G
B0027-100G
TCB0027-025GEA
36.34
CAD
TCB0027-025G
TCB0027-500G
TCB0027-100G
Sodium benzenesulfinate dihydrate ≥98.0% (by titrimetric analysis)
Sodium benzenesulphinate dihydrate
CAS Number: 25932-11-0
MDL Number: MFCD00149638
Molecular Formula: C6H6O2S
Molecular Weight: 164.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Benzenesulfinic Acid Sodium Salt
Formula:
C₆H₅NaO₂S·2H₂O |
MDL Number:
MFCD00149638 CAS Number: 25932-11-0 EINECS: 212-842-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...