Disodium benzene-1,3-disulfonate 55.0% (by HPLC)

Supplier: TCI America

B0023-25G B0023-500G
TCB0023-025GEA 25.72 CAD
TCB0023-025G TCB0023-500G
Disodium benzene-1,3-disulfonate 55.0% (by HPLC)
Disodium benzene-1,3-disulphonate

CAS Number: 831-59-4
MDL Number: MFCD00007492
Molecular Formula: C6H6O6S2
Molecular Weight: 282.19
Purity/Analysis Method: >55.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 1,3-Benzenedisulfonic Acid Disodium Salt

Formula: C₆H₄Na₂O₆S₂
MDL Number: MFCD00007492
CAS Number: 831-59-4
EINECS: 212-606-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1504f87bf2966ae6b3d960e96107ca4440bee38&vid=d1504f87bf2966ae6b3d960e96107ca4440bee38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 55.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR