Benzamide ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Benzoic acid amide
B0012-500G
B0012-25G
TCB0012-500GEA
322.32
CAD
TCB0012-500G
TCB0012-025G
Benzamide ≥99.0%
Benzamide
CAS Number: 55-21-0
MDL Number: MFCD00007968
Molecular Formula: C7H7NO
Molecular Weight: 121.14
Purity/Analysis Method: >99.0% (N)
Form: Crystal
Boiling point (°C): 288
Melting point (°C): 128
Flash Point (°C): 180
Other Product Names / Synonyms: Benzoylamide
Formula:
C₇H₇NO MW: 121.14 g/mol Boiling Pt: 288 Melting Pt: 124 Density: 1.35 g/cm³ (20 °C) Flash Pt: 180 Storage Temperature: Ambient |
MDL Number:
MFCD00007968 CAS Number: 55-21-0 EINECS: 200-227-7 Merck Index: 13,01059 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...