4-Chloro-7-nitrobenzofurazan ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A5592-1G A5592-5G
TCA5592-001GEA 67.94 CAD
TCA5592-001G TCA5592-005G
4-Chloro-7-nitrobenzofurazan ≥98.0% (by titrimetric analysis)
4-Chloro-7-nitrobenzofurazan

[for HPLC Labeling]
CAS Number: 10199-89-0
MDL Number: MFCD00005808
Molecular Formula: C6H2ClN3O3
Molecular Weight: 199.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 98


Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Chloro-7-nitrobenzofurazan, 4-Chloro-7-nitro-2,1,3-benzoxadiazole

Formula: C₆H₂ClN₃O₃
Melting Pt: 98 °C
MDL Number: MFCD00005808
CAS Number: 10199-89-0
EINECS: 233-496-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db17112959e70822706552a1d259f774c215e18&vid=50e95105022273f5da6a4281b265af843dad8b5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR