Luminol (3-Aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A5301-25G A5301-1G
TCA5301-025GEA 302.41 CAD
TCA5301-025G TCA5301-001G
Luminol (3-Aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis)
Luminol (3-Aminophthalhydrazide)

[Chemiluminescence Reagent]
CAS Number: 521-31-3
MDL Number: MFCD00006890
Molecular Formula: C8H7N3O2
Molecular Weight: 177.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 320


Product Categories: Bioscience, ROS/RNS Detection Reagents, Redox Research Reagents, Reagents for Nitric Oxide Research, Analytical Science, Luminols (Chemiluminescence Reagents)
Other Product Names / Synonyms: 3-Aminophthalhydrazide, 3-Aminophthaloylhydrazine, 5-Amino-2,3-dihydro-1,4-phthalazinedione

Formula: C₈H₇N₃O₂
Melting Pt: 320 °C
MDL Number: MFCD00006890
CAS Number: 521-31-3
EINECS: 208-309-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=febe78dd156ede1988f486774994196fcf081c0d&vid=8faeff09cdafbd0be4f36cf2ccb19b2695ce80fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR