Potassium tetrakis(4-chlorophenyl)borate ≥98.0%

Supplier: TCI America

Synonyms: Potassiumtetrakis(4-chlorophenyl)borate

A5132-5G A5132-1G
TCA5132-5GEA 783.68 CAD
TCA5132-5G TCA5132-001G
Potassium tetrakis(4-chlorophenyl)borate ≥98.0%
Potassium tetrakis(4-chlorophenyl)borate

[Anion for the neutral carrier type ion electrode]
CAS Number: 14680-77-4
MDL Number: MFCD00043105
Molecular Formula: C24H16BCl4K
Molecular Weight: 496.10
Purity/Analysis Method: >98.0% (HPLC,W)
Form: Crystal


Product Categories: Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Tetrakis(4-chlorophenyl)boron Potassium

Formula: C₂₄H₁₆BCl₄K
MDL Number: MFCD00043105
CAS Number: 14680-77-4
EINECS: 238-723-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b87f93b0bacd257d77f1565a1dc381340e64dbc5&vid=e37bc09b7cb0d65547bb578e57239623bcac5f6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,W)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR