meso-Tetraphenylporphine

Supplier: TCI America

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP

A5012-25G A5012-1G
TCA5012-025GEA 1413.31 CAD
TCA5012-025G TCA5012-001G
meso-Tetraphenylporphine
meso-Tetraphenylporphine

[Ultra-high sensitive spectrophotometric reagent for Cu]
CAS Number: 917-23-7
MDL Number: MFCD00011680
Molecular Formula: C44H30N4
Molecular Weight: 614.75
Form: Crystal
Color: Purple


Other Product Names / Synonyms: Tetraphenylporphyrin

Formula: C₄₄H₃₀N₄
MW: 614.75 g/mol
Density: 1.276 g/cm³ (20 °C)
MDL Number: MFCD00011680
CAS Number: 917-23-7
EINECS: 213-025-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5beb9473f042f6c9ea8a017f4f33fd8b79f90e&vid=958cdff18fe91aa8cd5fe3199fa8df986427f5e8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR