1-(2-Pyridylazo)-2-naphthol (PAN) 98,0
Supplier: TCI America
Synonyms:
PAN, 1-(2-Pyridylazo)-2-naphthol
A5000-1G
A5000-5G
TCA5000-001GEA
46.29
CAD
TCA5000-001G
TCA5000-005G
1-(2-Pyridylazo)-2-naphthol (PAN) 98,0
1-(2-Pyridylazo)-2-naphthol
[Metal indicator and spectrophotometric reagent for transition metals]
CAS Number: 85-85-8
MDL Number: MFCD00004071
Molecular Formula: C15H11N3O
Molecular Weight: 249.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 142
Product Categories: Analytical Science, Azo Compounds (Chelating Agents), Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 1-(2-Pyridylazo)-2-naphthol
Formula:
C₁₅H₁₁N₃O MW: 249.27 g/mol Boiling Pt: 412 Melting Pt: 142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004071 CAS Number: 85-85-8 EINECS: 201-637-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...