2-Amino-4-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2168-200MG A2168-1G A2168-5G
TCA2168-200MGEA 30.02 CAD
TCA2168-200MG TCA2168-1G TCA2168-5G
2-Amino-4-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-(trifluoromethyl)pyridine

CAS Number: 106447-97-6
MDL Number: MFCD00077455
Molecular Formula: C6H5F3N2
Molecular Weight: 162.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 73

Formula: C₆H₅F₃N₂
Melting Pt: 73 °C
MDL Number: MFCD00077455
CAS Number: 106447-97-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f1bc11b554d1b99a39413cb88b1637e32067ba0&vid=0f1bc11b554d1b99a39413cb88b1637e32067ba0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR