5-Amino-2-chlorobenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

A1913-25G A1913-250G
TCA1913-25GEA 312.84 CAD
TCA1913-25G TCA1913-250G
5-Amino-2-chlorobenzoic acid ≥98.0% (by HPLC, titration analysis)
5-Amino-2-chlorobenzoic acid

CAS Number: 89-54-3
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 188

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330
Melting Pt: 188
Density: 1.519 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007845
CAS Number: 89-54-3
EINECS: 201-916-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d9c72938e30e79a1f2aa58be1bed1329b6d501d&vid=c79e26a13052aaa169e8127d834af945fcfcd7b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR