2-Amino-5-phenylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1667-100MG A1667-1G
TCA1667-100MGEA 76 CAD
TCA1667-100MG TCA1667-1G
2-Amino-5-phenylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-phenylpyridine

CAS Number: 33421-40-8
MDL Number: MFCD01692452
Molecular Formula: C11H10N2
Molecular Weight: 170.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 135

Formula: C₁₁H₁₀N₂
Melting Pt: 135 °C
MDL Number: MFCD01692452
CAS Number: 33421-40-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9bdbe0df3afbbaca3d08863c4bbc4eb9ea86d5&vid=9f9bdbe0df3afbbaca3d08863c4bbc4eb9ea86d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR