2-Amino-5-bromopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Bromo-2-pyridylamine, 5-Bromopyridin-2-amine

A1587-250G A1587-25G
TCA1587-250GEA 227.52 CAD
TCA1587-250G TCA1587-25G
2-Amino-5-bromopyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-bromopyridine

CAS Number: 1072-97-5
MDL Number: MFCD00006323
Molecular Formula: C5H5BrN2
Molecular Weight: 173.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5818ca204b0255000f252738145346b8cca104e&vid=b5818ca204b0255000f252738145346b8cca104e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR