Dimethyl-5-aminoisophthalate ≥98.0%

Supplier: TCI America

Synonyms: Dimethyl 5-aminoisophthalate, 5-Aminoisophthalic acid dimethyl ester

A1586-25G A1586-250G
TCA1586-25GEA 63.2 CAD
TCA1586-25G TCA1586-250G
Dimethyl-5-aminoisophthalate ≥98.0%
Dimethyl 5-aminoisophthalate

CAS Number: 99-27-4
MDL Number: MFCD00008435
Molecular Formula: C10H11NO4
Molecular Weight: 209.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 181


Other Product Names / Synonyms: 5-Aminoisophthalic Acid Dimethyl Ester, Dimethyl 5-Aminobenzene-1,3-dicarboxylate

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00008435
CAS Number: 99-27-4
EINECS: 202-743-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa8d0d1d0ceea6203ced7293e8fa10949bb94eac&vid=fa8d0d1d0ceea6203ced7293e8fa10949bb94eac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR