2-Methoxyethyl acetoacetate ≥95.0%

Supplier: TCI America

A1583-500G A1583-25G
TCA1583-500GEA 186.44 CAD
TCA1583-500G TCA1583-25G
2-Methoxyethyl acetoacetate ≥95.0%
2-Methoxyethyl acetoacetate

CAS Number: 22502-03-0
MDL Number: MFCD00009648
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 103
Specific Gravity (20/20): 1.09


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Acetoacetic Acid 2-Methoxyethyl Ester

Formula: C₇H₁₂O₄
Boiling Pt: 120 °C
Density: 1.09 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00009648
CAS Number: 22502-03-0
EINECS: 245-043-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0207172fe2c1a652adc76e076fa3112cb47724e7&vid=0207172fe2c1a652adc76e076fa3112cb47724e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR