5-Amino-2-methylindole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1539-5G A1539-1G
TCA1539-005GEA 246.48 CAD
TCA1539-005G TCA1539-001G
5-Amino-2-methylindole ≥98.0% (by GC, titration analysis)
5-Amino-2-methylindole

CAS Number: 7570-49-2
MDL Number: MFCD02093426
Molecular Formula: C9H10N2
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C₉H₁₀N₂
Melting Pt: 158…159 °C
MDL Number: MFCD02093426
CAS Number: 7570-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edde7b1b7b01cc6c913ed95d3edfa62ab620f3cb&vid=edde7b1b7b01cc6c913ed95d3edfa62ab620f3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR