2-Amino-6-bromopyridine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A1529-25G
A1529-5G
TCA1529-025GEA
263.86
CAD
TCA1529-025G
TCA1529-005G
2-Amino-6-bromopyridine ≥98.0% (by titrimetric analysis)
2-Amino-6-bromopyridine
CAS Number: 19798-81-3
MDL Number: MFCD00137843
Molecular Formula: C5H5BrN2
Molecular Weight: 173.01
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 89
Formula:
C₅H₅BrN₂ Melting Pt: 89 °C |
MDL Number:
MFCD00137843 CAS Number: 19798-81-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...