L-allo-Threonine ≥99.0%
Supplier: TCI America
Synonyms:
H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH
A1519-1G
A1519-100MG
TCA1519-1GEA
720.48
CAD
TCA1519-1G
TCA1519-100MG
L-allo-Threonine ≥99.0%
L-allo-Threonine
CAS Number: 28954-12-3
MDL Number: MFCD00064268
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 10 deg (C=5, H2O)
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: L-allo-Threonine
Formula:
C₄H₉NO₃ MW: 119.12 g/mol |
MDL Number:
MFCD00064268 CAS Number: 28954-12-3 EINECS: 249-327-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...