D-allo-Threonine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

A1518-100MG A1518-1G
TCA1518-100MGEA 44.24 CAD
TCA1518-100MG TCA1518-1G
D-allo-Threonine ≥99.0% (by HPLC, titration analysis)
D-allo-Threonine

CAS Number: 24830-94-2
MDL Number: MFCD00004526
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -10 deg (C=5, H2O)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: D-allo-Threonine

Formula: C₄H₉NO₃
MDL Number: MFCD00004526
CAS Number: 24830-94-2
EINECS: 246-488-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e924b0fff4f5e2aa6218666d9250fb7eac7ec4c2&vid=e924b0fff4f5e2aa6218666d9250fb7eac7ec4c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR