2-Chloro-4-(trifluoromethyl)aniline ≥97.0%

Supplier: TCI America
Warning

A1511-25G A1511-5G
TCA1511-025GEA 179.65 CAD
TCA1511-025G TCA1511-5G
2-Chloro-4-(trifluoromethyl)aniline ≥97.0%
2-Chloro-4-(trifluoromethyl)aniline

CAS Number: 39885-50-2
MDL Number: MFCD00042563
Molecular Formula: C7H5ClF3N
Molecular Weight: 195.57
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 103
Specific Gravity (20/20): 1.43


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Chloro-4-trifluoromethylaniline

Formula: C₇H₅ClF₃N
Boiling Pt: 89 °C (0.5 mmHg)
Density: 1.43 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00042563
CAS Number: 39885-50-2
EINECS: 254-674-8
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ad37ebcd4bee6963a94707e71df04a6b03b11d4&vid=4ad37ebcd4bee6963a94707e71df04a6b03b11d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR