2-Chloro-6-(trifluoromethyl)aniline ≥97.0%

Supplier: TCI America

A1510-25G A1510-5G
TCA1510-025GEA 237 CAD
TCA1510-025G TCA1510-5G
2-Chloro-6-(trifluoromethyl)aniline ≥97.0%
2-Chloro-6-(trifluoromethyl)aniline

CAS Number: 433-94-3
MDL Number: MFCD00272565
Molecular Formula: C7H5ClF3N
Molecular Weight: 195.57
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 40
Flash Point (°C): 71
Specific Gravity (20/20): 1.42


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Chloro-6-trifluoromethylaniline

Formula: C₇H₅ClF₃N
Boiling Pt: 40 °C
Density: 1.42 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00272565
CAS Number: 433-94-3
EINECS: 207-091-8
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f03f97f1b24dd68d42c81b27e728e63d9f7b557&vid=1f03f97f1b24dd68d42c81b27e728e63d9f7b557&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR