2-Amino-3,5-dichloropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A1502-5G A1502-25G
TCA1502-005GEA 41.08 CAD
TCA1502-005G TCA1502-025G
2-Amino-3,5-dichloropyridine ≥98.0% (by GC, titration analysis)
2-Amino-3,5-dichloropyridine

CAS Number: 4214-74-8
MDL Number: MFCD00006313
Molecular Formula: C5H4Cl2N2
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 81

Formula: C₅H₄Cl₂N₂
Melting Pt: 81 °C
MDL Number: MFCD00006313
CAS Number: 4214-74-8
EINECS: 224-143-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f97d5964ffd01eb139f57126ea001a0b8df0ba75&vid=f97d5964ffd01eb139f57126ea001a0b8df0ba75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR