4-Amino-2-chlorophenol ≥98.0%

Supplier: TCI America

Synonyms: 3-Chloro-4-hydroxyaniline

A1496-5G A1496-25G
TCA1496-005GEA 49.82 CAD
TCA1496-005G TCA1496-25G
4-Amino-2-chlorophenol ≥98.0%
4-Amino-2-chlorophenol

CAS Number: 3964-52-1
MDL Number: MFCD00128891
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 153
Storage Temperature: 0-10°C

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00128891
CAS Number: 3964-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d5126639f909c71de4675e1982767c2f8f2f82a&vid=8d5126639f909c71de4675e1982767c2f8f2f82a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR