4-Acetoxybenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 4-(Ethanoyloxy)benzaldehyde, 4-Formylphenyl acetate

A1467-25G A1467-5G
TCA1467-25GEA 287.56 CAD
TCA1467-25G TCA1467-005G
4-Acetoxybenzaldehyde ≥98.0%
4-Acetoxybenzaldehyde

CAS Number: 878-00-2
MDL Number: MFCD00003384
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 265
Flash Point (°C): 110
Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Acetic Acid 4-Formylphenyl Ester, 4-Formylphenyl Acetate

Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 265 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00003384
CAS Number: 878-00-2
EINECS: 212-898-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=113278057db30ed4c37e5a1d602ed3c98ee13412&vid=113278057db30ed4c37e5a1d602ed3c98ee13412&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR