Isoquinoline-5-amine ≥98.0%

Supplier: TCI America

Synonyms: Isoquinolin-5-amine, 5-Aminoisoquinoline

A1465-5G A1465-1G
TCA1465-005GEA 77.42 CAD
TCA1465-005G TCA1465-001G
Isoquinoline-5-amine ≥98.0%
Isoquinoline-5-amine

CAS Number: 1125-60-6
MDL Number: MFCD00006907
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 130

Formula: C₉H₈N₂
Melting Pt: 130 °C
MDL Number: MFCD00006907
CAS Number: 1125-60-6
EINECS: 214-408-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2773218604e9f3f48eefab5d46c0939c51320be&vid=d2773218604e9f3f48eefab5d46c0939c51320be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR