Ethyl-3-aminobenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A1460-25G A1460-5G
TCA1460-025GEA 124.82 CAD
TCA1460-025G TCA1460-005G
Ethyl-3-aminobenzoate ≥98.0% (by GC, titration analysis)
Ethyl 3-aminobenzoate

CAS Number: 582-33-2
MDL Number: MFCD00007794
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 294
Flash Point (°C): 113
Freezing point (°C): 30


Other Product Names / Synonyms: 3-Aminobenzoic Acid Ethyl Ester

Formula: C₉H₁₁NO₂
Boiling Pt: 294 °C
Density: 1.173 g/cm³ (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00007794
CAS Number: 582-33-2
EINECS: 209-482-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fadc8a8203da50460c38a4e2c9a49866ac7e2bad&vid=fadc8a8203da50460c38a4e2c9a49866ac7e2bad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR