L-α-Aminoadipic acid ≥98.0%

Supplier: TCI America

Synonyms: L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid

A1457-1G A1457-5G
TCA1457-001GEA 239.69 CAD
TCA1457-001G TCA1457-005G
L-α-Aminoadipic acid ≥98.0%
L-α-Aminoadipic acid

CAS Number: 1118-90-7
MDL Number: MFCD00002636
Molecular Formula: C6H11NO4
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 25 deg (C=5, 6mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Aminohexanedioic Acid

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
MDL Number: MFCD00002636
CAS Number: 1118-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eea60ee19e49ca3f0597dbbac7de9f0e35c68acf&vid=eea60ee19e49ca3f0597dbbac7de9f0e35c68acf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR