3-Amino-1H-1,2,4-triazole-5-thiol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Amino-5-mercapto-1,2,4-triazole, 3-Amino-1,2,4-triazole-5-thiol, 3-Amino-4H-1,2,4-triazole-5-thiol, 3-Amino-5-mercapto-1,2,4-triazole 5-Amino-4H-1,2,4-triazole-3-thiol

A1455-250G A1455-5G A1455-25G
TCA1455-250GEA 412.38 CAD
TCA1455-250G TCA1455-005G TCA1455-025G
3-Amino-1H-1,2,4-triazole-5-thiol ≥98.0% (by titrimetric analysis)
3-Amino-1H-1,2,4-triazole-5-thiol

CAS Number: 16691-43-3
MDL Number: MFCD00005231
Molecular Formula: C2H4N4S
Molecular Weight: 116.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 3-Amino-4H-1,2,4-triazole-5-thiol

Formula: C₂H₄N₄S
MW: 116.15 g/mol
MDL Number: MFCD00005231
CAS Number: 16691-43-3
EINECS: 240-735-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=115cecce550a10cf1a2e20b39950349375edd2f4&vid=115cecce550a10cf1a2e20b39950349375edd2f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR