4-Amino-3-nitrophenol ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A1426-25G A1426-100G
TCA1426-025GEA 96.38 CAD
TCA1426-025G TCA1426-100G
4-Amino-3-nitrophenol ≥97.0% (by titrimetric analysis)
4-Amino-3-nitrophenol

CAS Number: 610-81-1
MDL Number: MFCD00066310
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 152


Other Product Names / Synonyms: 4-Hydroxy-2-nitroaniline

Formula: C₆H₆N₂O₃
Melting Pt: 152 °C
MDL Number: MFCD00066310
CAS Number: 610-81-1
EINECS: 210-236-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1106895bdd3abd7272b1bfc086c85f58939bbbd&vid=d1106895bdd3abd7272b1bfc086c85f58939bbbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR