Palladium(II) acetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Palladium diacetate, Palladium (II) acetate

A1424-5G A1424-1G
TCA1424-5GEA 1318.98 CAD
TCA1424-5G TCA1424-001G
Palladium(II) acetate ≥98.0% (by titrimetric analysis)
Palladium (II) acetate

CAS Number: 3375-31-3
MDL Number: MFCD00012453
Molecular Formula: C4H6O4Pd
Molecular Weight: 224.51
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Lambda max.: 400 nm (EtOH)


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Oxidation
Other Product Names / Synonyms: Acetic Acid Palladium(II) Salt

Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e1ff61fdcb945acd33cd2fb856fdbd08f9f6326&vid=2e1ff61fdcb945acd33cd2fb856fdbd08f9f6326&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR