5-Methylanthranil acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Amino-5-methylbenzoic acid, 6-Amino-m-toluic acid

A1421-25G A1421-5G
TCA1421-25GEA 86.9 CAD
TCA1421-25G TCA1421-005G
5-Methylanthranil acid ≥98.0%
5-Methylanthranil acid

CAS Number: 2941-78-8
MDL Number: MFCD00007909
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 6-Amino-m-toluic Acid, 5-Methylanthranilic Acid

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 317
Storage Temperature: Ambient
MDL Number: MFCD00007909
CAS Number: 2941-78-8
EINECS: 220-932-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9eb30b509099d9e458f0e34a7b7ca84d9b5dd4e&vid=5a349400ab15fbe03590630efcc8335fb6d8a85d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR