4-Amino-3-nitrobenzoic acid ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
A1417-25G
A1417-500G
A1417-100G
TCA1417-025GEA
94.8
CAD
TCA1417-025G
TCA1417-500G
TCA1417-100G
4-Amino-3-nitrobenzoic acid ≥95.0% (by HPLC, titration analysis)
4-Amino-3-nitrobenzoic acid
CAS Number: 1588-83-6
MDL Number: MFCD00017009
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Formula:
C₇H₆N₂O₄ Melting Pt: 290 °C |
MDL Number:
MFCD00017009 CAS Number: 1588-83-6 EINECS: 216-453-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...