2-Amino-6-chloropurine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A1407-1G A1407-5G
TCA1407-001GEA 50.56 CAD
TCA1407-001G TCA1407-005G
2-Amino-6-chloropurine ≥98.0% (by HPLC, titration analysis)
2-Amino-6-chloropurine

CAS Number: 10310-21-1
MDL Number: MFCD00075252
Molecular Formula: C5H4ClN5
Molecular Weight: 169.57
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₅H₄ClN₅
Melting Pt: 300 °C
MDL Number: MFCD00075252
CAS Number: 10310-21-1
EINECS: 233-686-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4bf2b9e0f616d98a54d88681c53703f87b35bb4&vid=f4bf2b9e0f616d98a54d88681c53703f87b35bb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR