3-Amino-o-toluic acid ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3-Amino-2-methylbenzoic acid
A1401-250G
A1401-25G
TCA1401-250GEA
383.94
CAD
TCA1401-250G
TCA1401-025G
3-Amino-o-toluic acid ≥99.0%
3-Amino-o-toluic acid
CAS Number: 52130-17-3
MDL Number: MFCD00075026
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 188
Other Product Names / Synonyms: 3-Amino-o-toluic Acid
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 347 Melting Pt: 181 Storage Temperature: Ambient |
MDL Number:
MFCD00075026 CAS Number: 52130-17-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...