5-Amino-4,6-dichloropyrimidine ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 4,6-Dichloropyrimidin-5-ylamine

A1395-5G A1395-25G A1395-1G
TCA1395-5GEA 62.68 CAD
TCA1395-5G TCA1395-25G TCA1395-001G
5-Amino-4,6-dichloropyrimidine ≥98.0% (by total nitrogen basis)
5-Amino-4,6-dichloropyrimidine

CAS Number: 5413-85-4
MDL Number: MFCD00006108
Molecular Formula: C4H3Cl2N3
Molecular Weight: 163.99
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 144

Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 144 °C
MDL Number: MFCD00006108
CAS Number: 5413-85-4
EINECS: 226-503-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80aa14359e8853901910a77339d409764d04447a&vid=80aa14359e8853901910a77339d409764d04447a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR