(R)-(-)-2-Aminobutyric acid 98,0

Supplier: TCI America

Synonyms: (R)-()-2-Aminobutyric acid, D-2-Aminobutyric acid, D(-)-2-Aminobutyric acid, H-D-Abu-OH

A1377-5G A1377-1G
TCA1377-005GEA 83.74 CAD
TCA1377-005G TCA1377-001G
(R)-(-)-2-Aminobutyric acid 98,0
(R)-(-)-2-Aminobutyric acid

CAS Number: 2623-91-8
MDL Number: MFCD00064414
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -21.0 deg (C=5, HCl(2+1))

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00064414
CAS Number: 2623-91-8
EINECS: 220-084-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4f6c62ba56159eb8026fa17d9329abd92802746&vid=c4f6c62ba56159eb8026fa17d9329abd92802746&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR