6-Aminoquinoline ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A1370-1G A1370-5G
TCA1370-001GEA 39.5 CAD
TCA1370-001G TCA1370-005G
6-Aminoquinoline ≥99.0% (by titrimetric analysis)
6-Aminoquinoline

CAS Number: 580-15-4
MDL Number: MFCD00006803
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 187
Melting point (°C): 116

Formula: C₉H₈N₂
Boiling Pt: 187 °C
Melting Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00006803
CAS Number: 580-15-4
EINECS: 209-453-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09791732cde54b0ed2d7e0aad11dc638efc943cb&vid=f9798de392e7962a0cc78dd4d015d0491d329da6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR