2-Amino-4,4,4-trifluorobutyric acid hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1367-100MG
A1367-1G
TCA1367-100MGEA
52.14
CAD
TCA1367-100MG
TCA1367-1G
2-Amino-4,4,4-trifluorobutyric acid hydrochloride ≥98.0% (by titrimetric analysis)
(±)-2-Amino-4,4,4-trifluorobutyric acid hydrochloride
CAS Number: 15959-93-0
MDL Number: MFCD00671488
Molecular Formula: C4H6F3NO2
Molecular Weight: 193.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₄H₇ClF₃NO₂ |
MDL Number:
MFCD00671488 CAS Number: 15959-93-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...