4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) ≥98.0% free radical

Supplier: TCI America

Synonyms: 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-Acetamido-2,2,6,6-Tetramethyl-N-Piperidinyloxy

A1348-25G A1348-5G
TCA1348-25GEA 217.95 CAD
TCA1348-25G TCA1348-005G
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) ≥98.0% free radical
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-Acetamido-TEMPO, free radical)

[Catalyst for Oxidation]
CAS Number: 14691-89-5
MDL Number: MFCD00043593
Molecular Formula: C11H21N2O2
Molecular Weight: 213.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 143
Flash Point (°C): 198


Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Acetamido-TEMPO

Formula: C₁₁H₂₁N₂O₂
MW: 213.3 g/mol
Melting Pt: 143 °C
Flash Pt: 198 °C
MDL Number: MFCD00043593
CAS Number: 14691-89-5
EINECS: 423-840-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15dbb5cbf66e41a91a8aa8ad29caeebdf827d9ce&vid=12c964cb56437ac9d80569752c7443d3ac80e387&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR