1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) ≥98.0%

Supplier: TCI America

Synonyms: 1,4,7,10,13-Pentaoxa-16-azacyclooctadecane, 1-Aza-18-crown 6-Ether, 1-Aza-18-crown-6

A1324-5G A1324-1G
TCA1324-5GEA 1714.3 CAD
TCA1324-5G TCA1324-001G
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) ≥98.0%
1-Aza-18-crown-6 (1,4,7,10,13-Pentaoxa-16-azacyclooctadecane)

CAS Number: 33941-15-0
MDL Number: MFCD00075466
Molecular Formula: C12H25NO5
Molecular Weight: 263.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 110


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10,13-Pentaoxa-16-azacyclooctadecane

Formula: C₁₂H₂₅NO₅
MW: 263.33 g/mol
Melting Pt: 51 °C
Flash Pt: 110 °C
MDL Number: MFCD00075466
CAS Number: 33941-15-0
EINECS: 251-751-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6278d0831a3277040c072485f02e2a4c55ea5ee5&vid=6278d0831a3277040c072485f02e2a4c55ea5ee5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR