1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) 98,0

Supplier: TCI America

Synonyms: 1,4,7,10-Tetraoxa-13-azacyclopentadecane, 1-Aza-15-crown 5-Ether, 1,4,7,10-Tetraoxa-13-azacyclopentadecane

A1323-5G A1323-1G
TCA1323-5GEA 737.86 CAD
TCA1323-5G TCA1323-001G
1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) 98,0
1-Aza-15-crown-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecane)

CAS Number: 66943-05-3
MDL Number: MFCD00075465
Molecular Formula: C10H21NO4
Molecular Weight: 219.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10-Tetraoxa-13-azacyclopentadecane

Formula: C₁₀H₂₁NO₄
MW: 219.28 g/mol
Melting Pt: 46 °C
Flash Pt: 113 °C
MDL Number: MFCD00075465
CAS Number: 66943-05-3
EINECS: 266-523-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82dbbca97cdde79616ab586173915c659c83c98b&vid=82dbbca97cdde79616ab586173915c659c83c98b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR