2-Amino-5-nitrobenzonitrile ≥98.0%
Supplier: TCI America
A1314-25G
A1314-250G
TCA1314-025GEA
97.96
CAD
TCA1314-025G
TCA1314-250G
2-Amino-5-nitrobenzonitrile ≥98.0%
2-Amino-5-nitrobenzonitrile
CAS Number: 17420-30-3
MDL Number: MFCD00007362
Molecular Formula: C7H5N3O2
Molecular Weight: 163.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 210
Flash Point (°C): 234
Other Product Names / Synonyms: 2-Cyano-4-nitroaniline, 5-Nitroanthranilonitrile
Formula:
C₇H₅N₃O₂ Melting Pt: 210 °C Flash Pt: 234 °C |
MDL Number:
MFCD00007362 CAS Number: 17420-30-3 EINECS: 241-446-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...