5-Aminoisophthalic acid 98,0

Supplier: TCI America
Warning

A1290-25G A1290-500G A1290-100G
TCA1290-025GEA 49.8 CAD
TCA1290-025G TCA1290-500G TCA1290-100G
5-Aminoisophthalic acid 98,0
5-Aminoisophthalic acid

CAS Number: 99-31-0
MDL Number: MFCD00002522
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 515
Melting Pt: 150…156 °C
MDL Number: MFCD00002522
CAS Number: 99-31-0
EINECS: 202-748-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cd8a8df3c5ce92d2b128d97c32dd6dba589fc61&vid=5cd8a8df3c5ce92d2b128d97c32dd6dba589fc61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR