2-Acetylthiazole ≥98.0%

Supplier: TCI America

Synonyms: 1-(Thiazol-2-yl)-1-ethanone

A1265-5G A1265-25G
TCA1265-005GEA 101.12 CAD
TCA1265-005G TCA1265-025G
2-Acetylthiazole ≥98.0%
2-Acetylthiazole

CAS Number: 24295-03-2
MDL Number: MFCD00005324
Molecular Formula: C5H5NOS
Molecular Weight: 127.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 78
Specific Gravity (20/20): 1.23

Formula: C₅H₅NOS
MW: 127.17 g/mol
Boiling Pt: 90 °C
Melting Pt: 65.5 °C
Density: 1.23 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00005324
CAS Number: 24295-03-2
EINECS: 246-134-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0eb830fdc1d699bfc6b934db562d15cf29038a0&vid=c0eb830fdc1d699bfc6b934db562d15cf29038a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR