5-Bromoanthranilic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Amino-5-bromobenzoic acid
A1254-5G
A1254-25G
TCA1254-005GEA
41.08
CAD
TCA1254-005G
TCA1254-025G
5-Bromoanthranilic acid ≥98.0%
5-Bromoanthranilic acid
CAS Number: 5794-88-7
MDL Number: MFCD00007823
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 208
Other Product Names / Synonyms: 2-Amino-5-bromobenzoic Acid
Formula:
C₇H₆BrNO₂ Boiling Pt: 342 °C (1013 hPa) Melting Pt: 208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007823 CAS Number: 5794-88-7 EINECS: 227-338-3 UN: 2811 ADR: 6.1,III Merck Index: 13,01389 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...