4-Methoxybenzaldehyde dimethyl acetal ≥97.0%

Supplier: TCI America

A1247-500ML A1247-25ML
TCA1247-500MLEA 639.74 CAD
TCA1247-500ML TCA1247-025ML
4-Methoxybenzaldehyde dimethyl acetal ≥97.0%
4-Methoxybenzaldehyde dimethyl acetal

CAS Number: 2186-92-7
MDL Number: MFCD00036507
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 253
Flash Point (°C): 98
Specific Gravity (20/20): 1.07


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 4-Methoxybenzaldehyde Dimethyl Acetal, alpha,alpha,4-Trimethoxytoluene

Formula: C₁₀H₁₄O₃
Boiling Pt: 253 °C
Density: 1.07 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00036507
CAS Number: 2186-92-7
EINECS: 218-577-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49aea794f23324d6788ca24ef15af44f0052f863&vid=49aea794f23324d6788ca24ef15af44f0052f863&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR