Ethyl hydrogen adipate ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A1223-500G A1223-25G
TCA1223-500GEA 728.38 CAD
TCA1223-500G TCA1223-025G
Ethyl hydrogen adipate ≥97.0% (by GC, titration analysis)
Ethyl hydrogen adipate

CAS Number: 626-86-8
MDL Number: MFCD00004419
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >97.0% (GC,T)
Flash Point (°C): 110
Freezing point (°C): 28


Other Product Names / Synonyms: Adipic Acid Monoethyl Ester, Ethyl Hydrogen Adipate

Formula: C₈H₁₄O₄
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 27.4 °C
Density: 1.0622 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004419
CAS Number: 626-86-8
EINECS: 210-966-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64b360bb6f286f0aef487a7e624b6352e4604697&vid=64b360bb6f286f0aef487a7e624b6352e4604697&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR