2-Amino-6-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-Chlorobenzothiazol-2-ylamine, 6-Chloro-1,3-benzothiazol-2-amine

A1216-25G A1216-500G
TCA1216-025GEA 39.5 CAD
TCA1216-025G TCA1216-500G
2-Amino-6-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-6-chlorobenzothiazole

CAS Number: 95-24-9
MDL Number: MFCD00053557
Molecular Formula: C7H5ClN2S
Molecular Weight: 184.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199

Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 199 °C
MDL Number: MFCD00053557
CAS Number: 95-24-9
EINECS: 202-402-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40945141e60bdb671bc9ef662388b3de98d4fbee&vid=40945141e60bdb671bc9ef662388b3de98d4fbee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR